Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110008L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025373.1 | NP_079649.1 | 1 | 241 | ||
GenBank mRNA | AK045209.1 | - | 1 | 129 | ||
AK151976.1 | - | 1 | 241 | |||
AK152028.1 | - | 1 | 241 |