Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025557.1 | NP_079833.1 | 3 | 518 | ||
GenBank mRNA | AK002772.1 | - | 3 | 489 | ||
AK032164.1 | - | 3 | 488 | |||
AK075738.1 | - | 3 | 488 | |||
AY304481.1 | - | 3 | 489 | |||
BC116208.1 | - | 3 | 548 | |||
BC126965.1 | - | 3 | 548 |