Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081188.1 | NP_001074657.1 | 3 | 280 | ||
GenBank mRNA | AK011294.1 | - | 3 | 267 | ||
AK129057.1 | - | 3 | 232 | |||
BC039655.1 | - | 3 | 265 | |||
BC052656.1 | - | 3 | 262 | 30041585 | ||
BC094932.1 | - | 3 | 273 | 30008071 | ||
BC132363.1 | - | 3 | 295 | 40130640 | ||
BC145675.1 | - | 3 | 295 | 40130978 |