Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctcfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081387.2 | NP_001074856.1 | 9 | 1857 | ||
GenBank mRNA | BC151026.1 | - | 9 | 1732 | 9087939 | |
DQ153171.1 | - | 9 | 1857 |