Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167923.1 | NP_001161395.1 | 33 | 5914 | ||
GenBank mRNA | AM748257.1 | - | 26 | 2650 |