Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081090.1 | NP_001074559.1 | 3 | 890 | ||
GenBank mRNA | AK076024.1 | - | 3 | 884 | ||
AK132230.1 | - | 2 | 812 | |||
AK163125.1 | - | 2 | 733 | |||
BC010672.1 | - | 3 | 861 | |||
BC021330.1 | - | 3 | 909 | |||
BC055464.1 | - | 2 | 792 | |||
BC096029.1 | - | 3 | 887 | |||
BC150893.1 | - | 3 | 856 | 9087804 | ||
BC157979.1 | - | 3 | 856 | 9007299 |