Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099277.1 | NP_001092747.1 | 3 | 2368 | ||
GenBank mRNA | BC157962.1 | - | 3 | 2389 | 9007287 | |
DQ864732.1 | - | 3 | 2368 |