Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025691.1 | NP_079967.1 | 19 | 2326 | ||
GenBank mRNA | AK017869.1 | - | 5 | 882 | ||
BC058769.1 | - | 2 | 1074 | |||
BC089462.1 | - | 10 | 1334 |