Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172415.1 | NP_001165886.1 | 2 | 620 | ||
NM_004323.5 | NP_004314.5 | 2 | 620 | |||
GenBank mRNA | AF022224.1 | - | 2 | 578 | ||
AF116273.1 | - | 2 | 548 | |||
AK096038.1 | - | 3 | 383 | |||
AK222749.1 | - | 2 | 567 | |||
AK316577.1 | - | 2 | 546 | |||
BC001936.1 | - | 2 | 581 | 2823774 | ||
BC014774.1 | - | 2 | 430 | |||
KU177932.1 | - | 2 | 320 | |||
KU177933.1 | - | 2 | 320 | |||
U46917.1 | - | 2 | 580 | |||
Z35491.1 | - | 2 | 598 |