Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271017.1 | NP_001257946.1 | 6 | 2179 | ||
NM_025773.3 | NP_080049.2 | 6 | 2258 | |||
GenBank mRNA | AK082233.1 | - | 6 | 2207 | ||
AK153165.1 | - | 1 | 1672 |