Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271016.1 | NP_001257945.1 | 7 | 651 | ||
GenBank mRNA | AK050923.1 | - | 7 | 625 |