Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9030624G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256489.1 | NP_001243418.1 | 4 | 1919 | ||
GenBank mRNA | AK018572.1 | - | 4 | 1897 |
Gene Symbol: 2410018L13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC063063.1 | - | 4 | 1945 |
Gene Symbol: Gm4983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030401.1 | - | 1796 | 5362444 |
Gene Symbol: Gm5784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149188.1 | - | 2 | 1663 |