Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004326.3 | NP_004317.2 | 5 | 955 | ||
XM_005272971.4 | XP_005273028.1 | 5 | 960 | |||
XM_005277417.3 | XP_005277474.1 | 5 | 984 | |||
GenBank mRNA | BC116451.1 | - | 2 | 350 | 7961941 | |
CN281038.1 | - | 2 | 391 | |||
Y13620.1 | - | 5 | 1043 |