Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019205.1 | NP_062078.1 | 1, 2 | 118 | ||
GenBank mRNA | U96637.1 | - | 1, 2 | 84 | ||
Y08358.1 | - | 1, 2 | 118 |