Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myeov2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163425.1 | NP_001156897.1 | 3 | 234 | ||
GenBank mRNA | AA764377.1 | - | 51 | |||
AK003315.1 | - | 3 | 215 | |||
AK012198.1 | - | 3 | 207 | |||
AK152318.1 | - | 3 | 201 | |||
BC049759.1 | - | 3 | 234 | |||
BC147334.1 | - | 3 | 220 | 9056392 | ||
BC147335.1 | - | 3 | 220 | 9056395 | ||
BC147659.1 | - | 3 | 220 | 9087448 | ||
BC147669.1 | - | 3 | 220 | 9087458 | ||
BY631936.1 | - | 3 | 229 |