Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025924.2 | NP_080200.1 | 7 | 793 | ||
GenBank mRNA | AK011127.1 | - | 7 | 770 | ||
AK016053.1 | - | 7 | 756 | |||
AK145471.1 | - | 750 | ||||
BC116988.1 | - | 7 | 793 | 40126307 | ||
BC119035.1 | - | 7 | 793 | 8733784 |