Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205239.1 | NP_001192168.1 | 22 | 2242 | ||
NM_029922.3 | NP_084198.2 | 22 | 2182 | |||
GenBank mRNA | AK009530.1 | - | 10 | 799 | ||
AK014142.1 | - | 13 | 985 | |||
AK035349.1 | - | 21 | 2036 | |||
AK075876.1 | - | 10 | 801 | |||
AK076321.1 | - | 21 | 2036 | |||
BC030184.1 | - | 7 | 462 | |||
BC031387.1 | - | 19 | 1770 | 5038718 | ||
BC052064.1 | - | 22 | 2180 | |||
BC058266.1 | - | 12 | 1780 | |||
BC062096.1 | - | 21 | 2099 | 5716648 |