Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025584.2 | NP_079860.2 | 931 | |||
GenBank mRNA | AB122023.1 | - | 931 | |||
AK002762.1 | - | 842 | ||||
AK004342.1 | - | 933 | ||||
AK010597.1 | - | 915 | ||||
AK044051.1 | - | 921 | ||||
AY262355.1 | - | 904 | ||||
AY262356.1 | - | 899 | ||||
BC019482.1 | - | 929 | 4218841 |