Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700042G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099295.2 | NP_001092765.1 | 1 | 213 | ||
GenBank mRNA | AK006685.1 | - | 1 | 213 | ||
AK018884.1 | - | 1 | 192 | |||
BU946248.1 | - | 1 | 193 |