Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004331.2 | NP_004322.1 | 2 | 379 | ||
XM_005273617.3 | XP_005273674.1 | 2 | 283 | |||
GenBank mRNA | AB004788.1 | - | 2 | 300 | ||
AF060922.1 | - | 2 | 333 | |||
AF067396.1 | - | 2 | 337 | |||
AF079221.1 | - | 2 | 337 | |||
AF452712.2 | - | 2 | 352 | |||
AF536326.1 | - | 2 | 315 | |||
AK290213.1 | - | 2 | 295 | |||
AK300286.1 | - | 2 | 304 | |||
AK300727.1 | - | 2 | 318 | |||
AK315870.1 | - | 2 | 256 | |||
AK316315.1 | - | 2 | 283 | |||
AL132665.1 | - | 2 | 353 | |||
BC001559.1 | - | 2 | 279 | 3459327 | ||
BC009603.1 | - | 2 | 347 | 4134765 | ||
BT019500.1 | - | 2 | 255 | |||
BT019501.1 | - | 2 | 255 | |||
CD558918.1 | - | 2 | 488 | |||
CR456701.1 | - | 2 | 255 |