Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081064.1 | NP_001074533.1 | 24 | 10702 | ||
GenBank mRNA | AK122240.1 | - | 6 | 6371 | ||
AK136542.1 | - | 3 | 2832 |