Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sympk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026605.2 | NP_080881.2 | 14 | 2112 | ||
GenBank mRNA | AK019497.1 | - | 9 | 1588 | ||
AK146894.1 | - | 14 | 2093 | |||
AK155093.1 | - | 14 | 2113 | |||
AK155375.1 | - | 14 | 2067 | |||
AK165591.1 | - | 14 | 2091 | |||
AK168541.1 | - | 14 | 2093 | |||
AK168757.1 | - | 14 | 2093 | |||
AK172040.1 | - | 15 | 2141 | |||
AK215540.1 | - | 2 | 249 | |||
BC049852.1 | - | 14 | 2057 | 4163760 |