Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sympk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026605.2 | NP_080881.2 | 6 | 522 | ||
GenBank mRNA | AK146894.1 | - | 6 | 504 | ||
AK155093.1 | - | 6 | 523 | |||
AK155375.1 | - | 6 | 477 | |||
AK165591.1 | - | 6 | 501 | |||
AK168541.1 | - | 6 | 504 | |||
AK168757.1 | - | 6 | 504 | |||
AK172040.1 | - | 6 | 477 | |||
BC049852.1 | - | 6 | 467 | 4163760 |