Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019234.1 | NP_062107.1 | 2 | 235 | ||
XM_006236026.2 | XP_006236088.1 | 2 | 413 | |||
GenBank mRNA | X66845.1 | - | 2 | 235 |