Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019239.2 | NP_062112.2 | 2 | 365 | ||
XM_006242072.2 | XP_006242134.1 | 2 | 460 | |||
XM_006242073.2 | XP_006242135.1 | 2 | 935 | |||
XM_006242074.2 | XP_006242136.1 | 2 | 338 | |||
XM_006242077.2 | XP_006242139.1 | 2 | 824 | |||
GenBank mRNA | D10852.1 | - | 2 | 365 |