Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026774.2 | NP_081050.2 | 1 | 51 | ||
GenBank mRNA | AI552736.1 | - | 1 | 51 |