Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cthrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026778.2 | NP_081054.1 | 4 | 871 | ||
GenBank mRNA | AK003674.1 | - | 4 | 867 | ||
AK076498.1 | - | 4 | 875 | |||
AK151282.1 | - | 4 | 872 |