Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026818.2 | NP_081094.2 | 8 | 3996 | ||
GenBank mRNA | AK004006.1 | - | 1 | 658 |