Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026818.2 | NP_081094.2 | 5 | 742 | ||
GenBank mRNA | BC006187.1 | - | 5 | 402 |