Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163721.1 | NP_001157193.1 | 4 | 575 | ||
NM_001163722.1 | NP_001157194.1 | 4 | 501 | |||
GenBank mRNA | AK002541.1 | - | 3 | 375 | ||
AK004513.1 | - | 4 | 682 | |||
AK012228.1 | - | 4 | 426 | |||
AK028394.1 | - | 5 | 625 | |||
AK038662.1 | - | 4 | 2121 | |||
AK158109.1 | - | 2 | 2343 | |||
AK165247.1 | - | 4 | 473 | |||
AK167398.1 | - | 4 | 490 | |||
BC055936.1 | - | 3 | 357 | |||
BC055944.1 | - | 398 | ||||
BC080682.1 | - | 2 | 668 | |||
BC099522.1 | - | 4 | 812 |