Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033039.2 | NP_001028211.1 | 1 | 310 | ||
GenBank mRNA | AK076812.1 | - | 1 | 33 | ||
AK140290.1 | - | 1 | 311 | |||
BC141196.1 | - | 1 | 223 | 9055808 |