Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026861.2 | NP_081137.2 | 4 | 517 | ||
GenBank mRNA | AK035384.1 | - | 1 | 92 | ||
AK049109.1 | - | 4 | 466 | |||
AK158177.1 | - | 4 | 466 | |||
AK199665.1 | - | 2 | 204 | |||
BC024467.1 | - | 4 | 503 | 4975777 |