Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026861.2 | NP_081137.2 | 9 | 1283 | ||
GenBank mRNA | AK035384.1 | - | 6 | 858 | ||
AK049109.1 | - | 9 | 1232 | |||
AK158177.1 | - | 9 | 1232 | |||
AK159755.1 | - | 4 | 641 | |||
BC017635.1 | - | 3 | 445 | |||
BC024467.1 | - | 9 | 1269 | 4975777 |