Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026871.1 | NP_081147.1 | 5 | 458 | ||
GenBank mRNA | AF356874.1 | - | 5 | 463 | ||
AK004497.1 | - | 5 | 460 | |||
AK170135.1 | - | 4 | 1774 | |||
BC086940.1 | - | 5 | 458 | 6775318 |