Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019255.1 | NP_062128.1 | 2 | 286 | ||
GenBank mRNA | FQ215004.1 | - | 2 | 383 | ||
FQ215168.1 | - | 2 | 365 | |||
FQ215755.1 | - | 2 | 365 | |||
FQ216734.1 | - | 2 | 401 | |||
FQ224130.1 | - | 2 | 457 | |||
FQ224197.1 | - | 2 | 380 | |||
Y09453.1 | - | 2 | 286 |