Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026891.2 | NP_081167.2 | 26 | 3448 | ||
GenBank mRNA | AK005239.1 | - | 3 | 388 | ||
AK034144.1 | - | 11 | 1120 | |||
AK157271.1 | - | 26 | 3428 | |||
BC117555.1 | - | 24 | 2733 | 40098039 |