Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102563.1 | NP_001096033.1 | 4 | 2032 | ||
GenBank mRNA | AK005319.1 | - | 3 | 1020 | ||
AK158895.1 | - | 2 | 2483 | |||
BC053419.1 | - | 5 | 1548 |
Gene Symbol: Pagr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC054744.1 | - | 2321 |