Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163431.1 | NP_001156903.1 | 4 | 367 | ||
NM_026919.1 | NP_081195.1 | 4 | 338 | |||
GenBank mRNA | AK007351.1 | - | 4 | 323 | ||
AK145833.1 | - | 4 | 331 | |||
AV456553.1 | - | 4 | 309 | |||
BC024923.1 | - | 1 | 1329 | |||
BC058335.1 | - | 4 | 294 | |||
BC085316.1 | - | 3 | 964 |