Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026922.1 | NP_081198.1 | 16 | 1580 | ||
GenBank mRNA | AK220351.1 | - | 14 | 1165 | ||
BC042772.1 | - | 7 | 598 | |||
EF688288.1 | - | 16 | 1424 |