Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1810022K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099674.1 | NP_001093144.1 | 3 | 258 | ||
GenBank mRNA | AK007585.1 | - | 3 | 259 | ||
BC045157.1 | - | 3 | 245 | |||
BC055766.1 | - | 3 | 246 | |||
BC083104.1 | - | 3 | 257 | |||
BC100487.1 | - | 3 | 254 | |||
BC100488.1 | - | 3 | 242 | |||
BC115807.1 | - | 3 | 416 | 40106016 |