Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kmt2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026984.1 | NP_081260.1 | 16 | 2573 | ||
GenBank mRNA | AK020646.1 | - | 1 | 156 | ||
AK082403.1 | - | 1 | 458 | |||
AK148169.1 | - | 16 | 2558 | |||
BC064079.1 | - | 5 | 759 |