Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040396.2 | NP_001035486.2 | 2 | 296 | ||
GenBank mRNA | AK013022.1 | - | 2 | 260 | ||
AK032838.1 | - | 2 | 273 | |||
AK047958.1 | - | 2 | 274 | |||
AK146778.1 | - | 2 | 184 | |||
BC019970.1 | - | 2 | 244 | 4458171 | ||
BC038867.1 | - | 2 | 172 | 1547418 | ||
BC043068.1 | - | 182 | ||||
BC132453.1 | - | 2 | 262 | 40130730 | ||
BC138424.1 | - | 2 | 262 | 8861444 | ||
CJ133950.1 | - | 2 | 297 |