Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027050.1 | NP_081326.1 | 3 | 259 | ||
GenBank mRNA | AK005748.1 | - | 3 | 239 | ||
EU477371.1 | - | 3 | 182 |