Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700023F06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254724.2 | NP_001241653.1 | 10 | 1172 | ||
GenBank mRNA | AK006273.1 | - | 4 | 500 | ||
BC100507.1 | - | 8 | 1103 | 30917743 | ||
BY706354.1 | - | 4 | 502 |