Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027182.2 | NP_081458.1 | 13 | 2211 | ||
GenBank mRNA | AK010336.1 | - | 13 | 2173 | ||
AK142463.1 | - | 2184 | ||||
AK146877.1 | - | 13 | 2212 | |||
BC023834.1 | - | 7 | 1419 | |||
BC064315.1 | - | 13 | 2134 |