Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027201.1 | NP_081477.1 | 6 | 1061 | ||
GenBank mRNA | AK010407.1 | - | 5 | 847 | ||
BC065404.1 | - | 4 | 674 |