Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081137.1 | NP_001074606.1 | 11 | 1357 | ||
GenBank mRNA | EU937530.1 | - | 12 | 1420 |