Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027340.2 | NP_081616.1 | 10 | 1399 | ||
GenBank mRNA | AK154333.1 | - | 10 | 1400 |