Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027455.2 | NP_081731.1 | 6 | 1199 | ||
GenBank mRNA | AK017598.1 | - | 6 | 862 | ||
AK037197.1 | - | 6 | 852 | |||
AK045974.1 | - | 6 | 1200 | |||
AK148394.1 | - | 6 | 870 | |||
BC020023.1 | - | 5 | 770 | 3491756 | ||
BC151027.1 | - | 6 | 943 | 9087940 | ||
BC151029.1 | - | 6 | 943 | 9087942 |