Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027455.2 | NP_081731.1 | 2 | 586 | ||
GenBank mRNA | AK017598.1 | - | 2 | 249 | ||
AK037197.1 | - | 2 | 239 | |||
AK045974.1 | - | 2 | 587 | |||
AK148394.1 | - | 2 | 257 | |||
BC151027.1 | - | 2 | 330 | 9087940 | ||
BC151029.1 | - | 2 | 330 | 9087942 |